Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-iodoaniline, 98%, Thermo Scientific™

Catalog No. AC311930050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC311930050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC311930050 Supplier Thermo Scientific Chemicals Supplier No. 311930050

Chemical Identifiers

CAS 29632-74-4
Molecular Formula C6H5FIN
Molecular Weight (g/mol) 237.01
MDL Number MFCD00011738
InChI Key CUMTUBVTKOYYOU-UHFFFAOYSA-N
Synonym 2-fluoro-4-iodo-phenylamine, benzenamine, 2-fluoro-4-iodo, 2-fluoro-4-iodobenzenamine, 2-fluoro-4-iodo aniline, 2-fluoro-4-iodo-aniline, 2-fluoro-4-iodophenylamine, zlchem 543, pubchem1532, 2-fluoro4-iodoaniline, 2-fluoro-4-iodaniline
PubChem CID 185694
IUPAC Name 2-fluoro-4-iodoaniline
SMILES C1=CC(=C(C=C1I)F)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 51°C to 57°C
Color White to Brown
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula IC6H3(F)NH2
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 237.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Fluoro-4-iodoaniline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.