Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-iodoaniline, 99%

Catalog No. p-7026452
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 29632-74-4 | C6H5FIN | 237.016 g/mol

2-Fluoro-4-iodoaniline is widely used in the pharmaceutical, chemical and food industries as a fundamental building block to create a variety of important compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-4-iodoaniline is widely used in the pharmaceutical, chemical and food industries as a fundamental building block to create a variety of important compounds.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29632-74-4
Molecular Formula C6H5FIN
Molecular Weight (g/mol) 237.016
MDL Number MFCD00011738
InChI Key CUMTUBVTKOYYOU-UHFFFAOYSA-N
Synonym 2-fluoro-4-iodo-phenylamine, benzenamine, 2-fluoro-4-iodo, 2-fluoro-4-iodobenzenamine, 2-fluoro-4-iodo aniline, 2-fluoro-4-iodo-aniline, 2-fluoro-4-iodophenylamine, zlchem 543, pubchem1532, 2-fluoro4-iodoaniline, 2-fluoro-4-iodaniline
PubChem CID 185694
IUPAC Name 2-fluoro-4-iodoaniline
SMILES C1=CC(=C(C=C1I)F)N

Specifications

Melting Point 55°C to 57°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 2081100
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 237.02
Percent Purity 99%
Chemical Name or Material 2-Fluoro-4-iodoaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.