Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-methylaniline, 99%

Catalog No. AAA1659114
Click to view available options
Quantity:
5 g
25 g

CAS: 452-80-2 | C7H8FN | 125.15 g/mol

2-Fluoro-4-methylaniline is used in the preparation of 6-chloro-5-fluoroindole via Leimgruber-Batcho reaction. It is also used in the preparation of an (S)-amino alcohol, 2-amino-3-(2-fluoro-4-methylphenyl)-propan-1-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-4-methylaniline is used in the preparation of 6-chloro-5-fluoroindole via Leimgruber-Batcho reaction. It is also used in the preparation of an (S)-amino alcohol, 2-amino-3-(2-fluoro-4-methylphenyl)-propan-1-ol.

Solubility
Not miscible in water.

Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-80-2
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00040975
InChI Key ZQEXBVHABAJPHJ-UHFFFAOYSA-N
Synonym 2-fluoro-4-methyaniline, 2-fluoro-p-toluidine, 4-amino-3-fluorotoluene, p-toluidine, 2-fluoro, benzenamine, 2-fluoro-4-methyl, 2-fluoro-4-methyl aniline, 2-fluoro-4-methyl-aniline, 2-fluoro-4-methyl-phenylamine, 2-fluoro-4-methylphenylamine, benzenamine, 2-fluoro-4-methyl-9ci
PubChem CID 67984
IUPAC Name 2-fluoro-4-methylaniline
SMILES CC1=CC=C(N)C(F)=C1

Specifications

Density 1.108
Boiling Point 70°C to 71°C (7 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.533
Quantity 25 g
UN Number UN2810
Beilstein 2637578
Solubility Information Not miscible in water.
Formula Weight 125.15
Percent Purity 99%
Chemical Name or Material 2-Fluoro-4-methylaniline
Show More Show Less
Hazard Category H227-H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H311-H373-H227-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
RTECSNumber XU6650000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.