Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-nitroaniline, 95%

Catalog No. p-7036782
Click to view available options
Quantity:
5 g
25 g

CAS: 369-35-7 | C6H5FN2O2 | 156.116 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Recommended storage temperature: 2 - 4°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 369-35-7
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.116
MDL Number MFCD00034560
InChI Key LETNCFZQCNCACQ-UHFFFAOYSA-N
Synonym 2-fluoro-4-nitrobenzenamine, 2-fluoro-4-nitro-phenylamine, benzenamine, 2-fluoro-4-nitro, 2-fluoro-4-nitrophenylamine, 4-amino-3-fluoronitrobenzene, 2-fluoro-4-nitroaniline, pubchem8503, acmc-1bncp, 2-fluoro-4-niroaniline, 2-fluoro-4-nitro-aniline
PubChem CID 101254
IUPAC Name 2-fluoro-4-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])F)N

Specifications

Melting Point 122°C to 130°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 156.12
Percent Purity 95%
Chemical Name or Material 2-Fluoro-4-nitroaniline
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 206-719-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.