Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-nitrotoluene 97.0+%, TCI America™
SDP

Catalog No. F029225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F029225G 25 g
F02925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F029225G Supplier TCI America Supplier No. F029225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1427-07-2
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD00007199
InChI Key WIQISTBTOQNVCE-UHFFFAOYSA-N
Synonym 2-fluoro-4-nitrotoluene, toluene, 2-fluoro-4-nitro, benzene, 2-fluoro-1-methyl-4-nitro, 3-fluoro-4-methylnitrobenzene, 1-methyl-2-fluoro-4-nitrobenzene, 2-fluoro-1-methyl-4-nitro-benzene, pubchem1592, 2-fluoro4-nitrotoluene, acmc-209cov, 2-fluor-4-nitro-toluol
PubChem CID 74025
IUPAC Name 2-fluoro-1-methyl-4-nitrobenzene
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])F

Specifications

Color Yellow
Quantity 25 g
Formula Weight 155.13
Percent Purity ≥97.0% (GC)
Physical Form Crystals
Chemical Name or Material 2-Fluoro-4-nitrotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.