Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-4-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F08165G
Click to view available options
Quantity:
1 g
5 g

Specifications

CAS 503309-11-3
Melting Point 116°C
Color White-Yellow
Molecular Formula C7H5BF4O2
MDL Number MFCD03094995
Quantity 5 g
Synonym 2-fluoro-4-trifluoromethyl phenylboronic acid, 2-fluoro-4-trifluoromethylphenylboronic acid, 2-fluoro-4-trifluoromethyl benzeneboronic acid, 2-fluoro-4-trifluoromethyl phenyl boronic acid, boronic acid, 2-fluoro-4-trifluoromethyl phenyl
InChI Key SSFSVKVWAURAAM-UHFFFAOYSA-N
SMILES B(C1=C(C=C(C=C1)C(F)(F)F)F)(O)O
IUPAC Name [2-fluoro-4-(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 207.919
PubChem CID 16414145
Formula Weight 207.92
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.