Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-hydroxypyridine 98.0+%, TCI America™
SDP

Catalog No. F09805G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 55758-32-2
Molecular Formula C5H4FNO
Molecular Weight (g/mol) 113.091
MDL Number MFCD01075040
InChI Key HTRLNWYWOKWCLV-UHFFFAOYSA-N
Synonym 2-fluoro-5-hydroxypyridine, 6-fluoro-3-pyridinol, 6-fluoro-3-hydroxypyridine, 3-pyridinol, 6-fluoro, pubchem6321, 6-fluoropyridine-3-ol, 6-fluoro-pyridin-3-ol, acmc-1b21k, 3-hydroxy-6-fluoro-pyridine, ksc490s4n
PubChem CID 2763108
IUPAC Name 6-fluoropyridin-3-ol
SMILES C1=CC(=NC=C1O)F

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 5 g
Formula Weight 113.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-5-hydroxypyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.