Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-iodobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. F116125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F116125G 25 g
F11615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F116125G Supplier TCI America Supplier No. F116125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59382-39-7
Molecular Formula C7H3F4I
Molecular Weight (g/mol) 290.00
MDL Number MFCD00236678
InChI Key DKLKYTATXLQGMX-UHFFFAOYSA-N
Synonym 1-Fluoro-4-iodo-2-(trifluoromethyl)benzene
PubChem CID 2737344
IUPAC Name 1-fluoro-4-iodo-2-(trifluoromethyl)benzene
SMILES FC1=C(C=C(I)C=C1)C(F)(F)F

Specifications

Color Yellow
Quantity 25 g
Formula Weight 290.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-iodobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.