Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Fluoro-5-iodopyridine, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
Specifications
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 35°C to 36°C |
Color | White to Yellow |
Flash Point | 95°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Quantity | 5 g |
Formula Weight | 222.99 |
Percent Purity | 97% |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Harmful if swallowed. Causes skin irritation. May cause an allergic skin reaction. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/e |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction