Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-iodotoluene 98.0+%, TCI America™
SDP

Catalog No. F100425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F100425G 25 g
F10045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F100425G Supplier TCI America Supplier No. F100425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 452-68-6
Molecular Formula C7H6FI
Molecular Weight (g/mol) 236.028
MDL Number MFCD00013710
InChI Key DYQIYXDSKUUZRI-UHFFFAOYSA-N
Synonym 2-fluoro-5-iodotoluene, benzene, 1-fluoro-4-iodo-2-methyl, 1-fluoro-4-iodo-2-methyl-benzene, pubchem1619, 2-fluoro-5-iodootoluene, acmc-20a0l4, ksc493m8f, 4-fluoro-3-methyl-iodobenzene, buttpark 45\01-60
PubChem CID 521178
IUPAC Name 1-fluoro-4-iodo-2-methylbenzene
SMILES CC1=C(C=CC(=C1)I)F

Specifications

Color Yellow
Boiling Point 87°C
Quantity 25 g
Formula Weight 236.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-iodotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.