Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-5-nitrobenzaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Plastic bottle
27996-87-8
C7H4FNO3
169.11
MFCD00042298
VVXFDFQEIRGULC-UHFFFAOYSA-N
fluoro nitrobenzaldehyde,5-nitro-2-fluorobenzaldehyde,4-fluoro-3-formylnitrobenzene,2-fluoro-5-nitro-benzaldehyde,benzaldehyde, 2-fluoro-5-nitro,pubchem1459,acmc-209h0h,akos bb460,asischem d51510,2-fluoro-5 nitrobenzaldehyde
2734770
2-fluoro-5-nitrobenzaldehyde
C1=CC(=C(C=C1[N+](=O)[O-])C=O)F
27996-87-8
C7H4FNO3
169.11
MFCD00042298
VVXFDFQEIRGULC-UHFFFAOYSA-N
fluoro nitrobenzaldehyde,5-nitro-2-fluorobenzaldehyde,4-fluoro-3-formylnitrobenzene,2-fluoro-5-nitro-benzaldehyde,benzaldehyde, 2-fluoro-5-nitro,pubchem1459,acmc-209h0h,akos bb460,asischem d51510,2-fluoro-5 nitrobenzaldehyde
2734770
2-fluoro-5-nitrobenzaldehyde
C1=CC(=C(C=C1[N+](=O)[O-])C=O)F
Melting Point 57°C to 60°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Color Beige to Brown
Quantity 1g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula FC6H3(NO2)CHO
Formula Weight 169.11
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-nitrobenzaldehyde
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning