Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-6-nitrotoluene, 98%, Thermo Scientific™

Catalog No. AC277230500
Click to view available options
Quantity:
25 g
50 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 769-10-8
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.13
MDL Number MFCD00007197
InChI Key GXPIVRKDWZKIKZ-UHFFFAOYSA-N
Synonym 2-fluoro-6-nitrotoluene, 1-fluor-2-methyl-3-nitrobenzene, 2-nitro-6-fluorotoluene, 6-fluoro-2-nitrotoluene, benzene, 1-fluoro-2-methyl-3-nitro, 3-fluoro-2-methyl-1-nitrobenzene, 1-fluor-2-methyl-3-nitrobenzol, pubchem1594, acmc-1bc1k
PubChem CID 69854
IUPAC Name 1-fluoro-2-methyl-3-nitrobenzene
SMILES CC1=C(F)C=CC=C1[N+]([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.2700g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5220 to 1.5240 (20°C, 589nm)
Linear Formula CH3C6H3(NO2)F
Quantity 50 g
Beilstein 05, 326
Formula Weight 155.13
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-6-nitrotoluene, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS P Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-203-3
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.