Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoro-N-(4-fluorobenzyl)aniline, 97%, Thermo Scientific™

Catalog No. AAH5647103
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5647103 1 g
AAH56471MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5647103 Supplier Thermo Scientific Chemicals Supplier No. H5647103

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
A novel class of potent CCR3 receptor antagonists were designed and synthesized from the derivative of this compound.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886791-73-7
Molecular Formula C13H11F2N
Molecular Weight (g/mol) 219.235
InChI Key KQBMUYYUEWLGQP-UHFFFAOYSA-N
Synonym 2-fluoro-n-4-fluorobenzyl aniline, 2-fluoro-n-4-fluorophenyl methyl aniline, 4-fluorobenzylamine, n-2-fluorophenyl
PubChem CID 6422135
IUPAC Name 2-fluoro-N-[(4-fluorophenyl)methyl]aniline
SMILES C1=CC=C(C(=C1)NCC2=CC=C(C=C2)F)F

Specifications

Quantity 1 g
Formula Weight 219.23
Percent Purity 97%
Chemical Name or Material 2-Fluoro-N-(4-fluorobenzyl)aniline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.