Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoroadenine 98.0+%, TCI America™
SDP

Catalog No. F06471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06471G 1 g
F0647200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06471G Supplier TCI America Supplier No. F06471G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 700-49-2
Molecular Formula C5H4FN5
Molecular Weight (g/mol) 153.12
MDL Number MFCD01632749
InChI Key WKMPTBDYDNUJLF-UHFFFAOYSA-N
Synonym 2-fluoroadenine, 2-fluoro-6-aminopurine, 2-fluoro-1h-purin-6-amine, 2-fad, 2-fluoro-9h-purin-6-amine, 6-amino-2-fluoropurine, 2-fluoro-7 9 h-purin-6-ylamine, purine, 6-amino-2-fluoro, fluoroadenine, 1h-purin-6-amine, 2-fluoro
PubChem CID 12790
ChEBI CHEBI:72457
IUPAC Name 2-fluoro-7H-purin-6-amine
SMILES NC1=C2NC=NC2=NC(F)=N1

Specifications

Quantity 1 g
Formula Weight 153.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoroadenine
RTECSNumber AU6264700
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.