Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoroaniline, 99+%

Catalog No. AC119271000
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle

CAS: 348-54-9 | C6H6FN | 111.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 348-54-9
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.12
MDL Number MFCD00007642
InChI Key FTZQXOJYPFINKJ-UHFFFAOYSA-N
Synonym o-fluoroaniline, benzenamine, 2-fluoro, 1-amino-2-fluorobenzene, 2-fluorobenzenamine, 2-fluoro-phenylamine, 2-fluoro-benzenamine, fluoroaniline-2, o-aminofluorobenzene, aniline, o-fluoro, 2-fluorophenylamine
PubChem CID 9584
ChEBI CHEBI:27526
IUPAC Name 2-fluoroaniline
SMILES NC1=CC=CC=C1F

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -29°C
Color Colorless
Density 1.1510g/mL
Boiling Point 182°C to 183°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5400 to 1.542
Linear Formula FC6H4NH2
Quantity 100 mL
Beilstein 12, I, 296
Specific Gravity 1.151
Solubility Information Solubility in water: 17g/L in water (20°C). Other solubilities: soluble in carbon tetrachloride,diethyl ether and,ethanol
Formula Weight 111.12
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Fluoroaniline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-478-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.