Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobenzaldehyde, 97%

Catalog No. AC119321000
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle

CAS: 446-52-6 | C7H5FO | 124.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-52-6
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.11
MDL Number MFCD00003302
InChI Key ZWDVQMVZZYIAHO-UHFFFAOYSA-N
Synonym o-fluorobenzaldehyde, benzaldehyde, 2-fluoro, benzaldehyde, o-fluoro, unii-7zx20dxb9z, 2-fluor-benzaldehyd, 2-fluoro-benzaldehyde, benzaldehyde, fluoro, ortho-fluorobenzaldehyde, 7zx20dxb9z, fluorobenzaldehyde
PubChem CID 67970
IUPAC Name 2-fluorobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -44.5°C
Color Brown to Colorless
Density 1.1700g/mL
Boiling Point 172°C to 174°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5205 to 1.5225
Linear Formula FC6H4CHO
Quantity 100 mL
Beilstein 07, I, 132
Specific Gravity 1.17
Solubility Information Solubility in water: insoluble
Formula Weight 124.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzaldehyde, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-171-2
RTECSNumber CU6140000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.