Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobenzaldehyde, 97%

Catalog No. AAA1380022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1380022 100 g
AAA1380006 5 g
AAA1380014 25 g
AAA1380036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1380022 Supplier Thermo Scientific Chemicals Supplier No. A1380022
Add to Cart

CAS: 446-52-6 | C7H5FO | 124.114 g/mol

2?-(2-Fluorobenzylidene)-2-hydroxybenzohydrazide was synthesized by the reaction of 2-hydroxybenzoylhydrazine with 2-fluorobenzaldehyde in ethanol. Synthesis of n-aryl indolines from 2-fluorobenzaldehyde dimethylhydrazone derivatives which is an approach to preparation of c(aryl)-n(amine) bond atropisomeric amines. The o-dialkylaminobenzaldehydes were conveniently prepared by nucleophilic displacement of the activated fluorine in 2-fluorobenzaldehyde with the required dialkylamine in hot dimethylformamide. N?-(2-Fluorobenzylidene)-2-(quinolin-8-yloxy) acetohydrazide methanol solvate in the reaction of quinolin-8-yloxyacetic acid hydrazide and 2-fluorobenzaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-(2-Fluorobenzylidene)-2-hydroxybenzohydrazide was synthesized by the reaction of 2-hydroxybenzoylhydrazine with 2-fluorobenzaldehyde in ethanol. Synthesis of n-aryl indolines from 2-fluorobenzaldehyde dimethylhydrazone derivatives which is an approach to preparation of c(aryl)-n(amine) bond atropisomeric amines. The o-dialkylaminobenzaldehydes were conveniently prepared by nucleophilic displacement of the activated fluorine in 2-fluorobenzaldehyde with the required dialkylamine in hot dimethylformamide. N′-(2-Fluorobenzylidene)-2-(quinolin-8-yloxy) acetohydrazide methanol solvate in the reaction of quinolin-8-yloxyacetic acid hydrazide and 2-fluorobenzaldehyde.

Solubility
Insoluble in water.

Notes
Air Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, reducing agents, air, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-52-6
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.114
MDL Number MFCD00003302
InChI Key ZWDVQMVZZYIAHO-UHFFFAOYSA-N
Synonym o-fluorobenzaldehyde, benzaldehyde, 2-fluoro, benzaldehyde, o-fluoro, unii-7zx20dxb9z, 2-fluor-benzaldehyd, 2-fluoro-benzaldehyde, benzaldehyde, fluoro, ortho-fluorobenzaldehyde, 7zx20dxb9z, fluorobenzaldehyde
PubChem CID 67970
IUPAC Name 2-fluorobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)F

Specifications

Melting Point -45°C
Density 1.18
Boiling Point 172°C to 174°C
Flash Point 55°C (131°F)
Odor Odorless
Refractive Index 1.522
Quantity 100 g
UN Number UN1989
Beilstein 507155
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 124.11
Percent Purity 97%
Chemical Name or Material 2-Fluorobenzaldehyde
Show More Show Less
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H226-H302-H315-H319-H335
Flammable liquid and vapour.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.
EINECSNumber 207-171-2
RTECSNumber CU6140000
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.