Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobenzoic acid, 98+%

Catalog No. p-7081137
Click to view available options
Quantity:
100 g
500 g

Catalog No. AAA1101522

Supplier: Thermo Scientific Chemicals A1101522

Only null left
Add to Cart

CAS: 445-29-4 | C7H5FO2 | 140.113 g/mol

2-Fluorobenzoic acid may be employed in the preparation of zaragozic acid A analogs. It is also used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluorobenzoic acid may be employed in the preparation of zaragozic acid A analogs. It is also used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 445-29-4
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.113
MDL Number MFCD00002405
InChI Key NSTREUWFTAOOKS-UHFFFAOYSA-N
Synonym o-fluorobenzoic acid, benzoic acid, 2-fluoro, ortho-fluorobenzoic acid, o-fluorbenzoesaeure, benzoic acid, o-fluoro, fluorobenzoic acid, unii-64uz32koo4, o-fluorbenzoesaeure german, ofba, 2-fluoro-benzoic acid
PubChem CID 9935
ChEBI CHEBI:19577
IUPAC Name 2-fluorobenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)F

Specifications

Melting Point 122°C to 125°C
Density 1.46
Quantity 100 g
Beilstein 971265
Solubility Information Slightly soluble in water.
Formula Weight 140.11
Percent Purity ≥98%
Chemical Name or Material 2-Fluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-158-1
RTECSNumber DH0250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.