Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobenzoic acid, 99%

Catalog No. p-133948
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle

CAS: 445-29-4 | C7H5FO2 | 140.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 445-29-4
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.11
MDL Number MFCD00002405
InChI Key NSTREUWFTAOOKS-UHFFFAOYSA-N
Synonym o-fluorobenzoic acid, benzoic acid, 2-fluoro, ortho-fluorobenzoic acid, o-fluorbenzoesaeure, benzoic acid, o-fluoro, fluorobenzoic acid, unii-64uz32koo4, o-fluorbenzoesaeure german, ofba, 2-fluoro-benzoic acid
PubChem CID 9935
ChEBI CHEBI:19577
IUPAC Name 2-fluorobenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 123°C to 125°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula FC6H4CO2H
Quantity 25 g
Beilstein 09, 333
Solubility Information Solubility in water: 7.2g/L (25°C)
Formula Weight 140.11
Percent Purity 99%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 2-Fluorobenzoic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 207-158-1
RTECSNumber DH0250000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.