Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobenzonitrile, 99%

Catalog No. p-3778608
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 394-47-8 | C7H4FN | 121.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 394-47-8
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.11
MDL Number MFCD00001773
InChI Key GDHXJNRAJRCGMX-UHFFFAOYSA-N
Synonym o-fluorobenzonitrile, benzonitrile, 2-fluoro, o-cyanofluorobenzene, 2-fluorobenzonitril, 2-cyanofluorobenzene, benzonitrile, o-fluoro, fluorobenzonitrile, 2-fluorobenzenecarbonitrile, 2-fluoro-benzonitrile, 1-cyano-2-fluorobenzene
PubChem CID 67855
IUPAC Name 2-fluorobenzonitrile
SMILES FC1=CC=CC=C1C#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Brown to Colorless
Density 1.1100g/mL
Boiling Point 90°C (21.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5063 to 1.5083
Quantity 5 g
Specific Gravity 1.11
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 121.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzonitrile
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fre
EINECSNumber 206-897-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.