Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorobenzyl alcohol, 98%, Thermo Scientific™

Catalog No. AC171970250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC171970250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC171970250 Supplier Thermo Scientific Chemicals Supplier No. 171970250

Chemical Identifiers

CAS 446-51-5
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00004601
InChI Key QEHXDOJPVIHUDO-UHFFFAOYSA-N
Synonym 2-fluorobenzyl alcohol, 2-fluorophenyl methanol, o-fluorobenzyl alcohol, benzenemethanol, 2-fluoro, 2-fluorobenzylalcohol, 2-fluoro-benzenemethanol, 2-fluorobenzylic alcohol, 2-fluorophenyl methan-1-ol, pubchem3485, acmc-1asq8
PubChem CID 67969
IUPAC Name (2-fluorophenyl)methanol
SMILES OCC1=CC=CC=C1F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1700g/mL
Boiling Point 200.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5126 to 1.5146
Linear Formula FC6H4CH2OH
Quantity 25 g
Beilstein 06, I, 222
Specific Gravity 1.17
Formula Weight 126.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzyl alcohol, 98%
Show More Show Less
EINECSNumber 207-170-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.