Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorocyclohexanone, 96%

Catalog No. AAH2603203
Click to view available options
Quantity:
250 mg
1 g

CAS: 694-82-6 | C6H9FO | 116.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-82-6
Molecular Formula C6H9FO
Molecular Weight (g/mol) 116.135
MDL Number MFCD01076476
InChI Key VQYOFTVCYSPHPG-UHFFFAOYSA-N
Synonym 2-fluorocyclohexanone, cyclohexanone, 2-fluoro, 2-fluorocylcohexanone, acmc-209o8v, 2-fluoro-cyclohexan-1-one
PubChem CID 11829404
IUPAC Name 2-fluorocyclohexan-1-one
SMILES C1CCC(=O)C(C1)F

Specifications

Density 1.08
Boiling Point 78°C (17 mmHg)
Flash Point 71°C (160°F)
Refractive Index 1.444
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 116.13
Percent Purity 96%
Chemical Name or Material 2-Fluorocyclohexanone
Hazard Category H227-H302-H319-H334
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P261-P264b-P270-P280-P285-P301+P312-P304+P341-P305+P351+P338-P330-P342+P311-P370+P378q-P501c
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.