Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluoroethanol, 95%

Catalog No. p-4812808
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

CAS: 371-62-0 | C2H5FO | 64.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371-62-0
Molecular Formula C2H5FO
Molecular Weight (g/mol) 64.06
InChI Key GGDYAKVUZMZKRV-UHFFFAOYSA-N
Synonym ethanol, 2-fluoro, fluoroethanol, 2-fluoro-1-ethanol, ethylene fluorohydrin, beta-fluoroethanol, 2-fluoroethan-1-ol, monofluorethanol, monofluoroethanol, 2-fluoro-ethanol, unii-4m427a5hml
PubChem CID 9737
IUPAC Name 2-fluoroethanol
SMILES C(CF)O

Specifications

Melting Point -26.5°C
Density 1.1080g/mL
Boiling Point 100.0°C to 104.0°C
Flash Point 34°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula FCH2CH2OH
Quantity 5 g
Beilstein 01,I,17
Specific Gravity 1.108
Solubility Information Solubility in water: miscible
Formula Weight 64.06
Percent Purity 95%
Chemical Name or Material 2-Fluoroethanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapour.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF
EINECSNumber 206-740-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.