Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorophenethylamine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
52721-69-4
C8H10FN
139.17
MFCD00075502
RIKUOLJPJNVTEP-UHFFFAOYSA-N
2-fluorophenethylamine,2-2-fluorophenyl ethanamine,2-2-fluorophenyl ethylamine,benzeneethanamine, 2-fluoro,2-2-fluoro-phenyl-ethylamine,2-2-fluorophenyl ethan-1-amine,2-fluoro-phenethylamine,2-fluorophenethyl amine,2-fluorobenzeneethanamine,benzeneethanamine,2-fluoro
643357
2-(2-fluorophenyl)ethanamine
C1=CC=C(C(=C1)CCN)F
52721-69-4
C8H10FN
139.17
MFCD00075502
RIKUOLJPJNVTEP-UHFFFAOYSA-N
2-fluorophenethylamine,2-2-fluorophenyl ethanamine,2-2-fluorophenyl ethylamine,benzeneethanamine, 2-fluoro,2-2-fluoro-phenyl-ethylamine,2-2-fluorophenyl ethan-1-amine,2-fluoro-phenethylamine,2-fluorophenethyl amine,2-fluorobenzeneethanamine,benzeneethanamine,2-fluoro
643357
2-(2-fluorophenyl)ethanamine
C1=CC=C(C(=C1)CCN)F
Boiling Point 64°C (0.6mmHg)
Flash Point 77°C
Infrared Spectrum Authentic
Specific Gravity 1.066
Density 1.0660g/mL
Refractive Index 1.5100 to 1.512
Packaging Glass bottle
Color Orange to Yellow
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula FC6H4CH2CH2NH2
Formula Weight 139.17
Physical Form Liquid
Percent Purity ≥96.0% (GC)
Chemical Name or Material 2-Fluorophenethylamine, 99%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

TSCA : TSCA