Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193354
Click to view available options
Quantity:
5 g
25 g

Specifications

CAS 1993-03-9
Melting Point 110°C
Molecular Formula C6H6BFO2
Quantity 5 g
MDL Number MFCD00674013
Synonym 2-fluorophenyl boronic acid, 2-fluorobenzeneboronic acid, 2-fluorophenyl boranediol, o-fluorophenylboronic acid, boronic acid, b-2-fluorophenyl
InChI Key QCSLIRFWJPOENV-UHFFFAOYSA-N
SMILES B(C1=CC=CC=C1F)(O)O
IUPAC Name (2-fluorophenyl)boronic acid
Molecular Weight (g/mol) 139.92
PubChem CID 2734354
Formula Weight 139.92
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.