Learn More
2-Fluoropropionic acid, 96%, Thermo Scientific Chemicals
Description
2-Fluoropropionic acid is used in the medical field for diagnostic purposes. It is labeled with 18F and is used as an imaging agent for detecting prostate cancer in humans. It is also used to prepare 2-fluoro fatty acids, compounds that exhibit antifungal properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Fluoropropionic acid is used in the medical field for diagnostic purposes. It is labeled with 18F and is used as an imaging agent for detecting prostate cancer in humans. It is also used to prepare 2-fluoro fatty acids, compounds that exhibit antifungal properties.
Solubility
Partly miscible in water.
Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents, strong bases.
Specifications
Specifications
| Density | 1.214 |
| Boiling Point | 66°C to 67°C (30 mmHg) |
| Flash Point | 82°C (180°F) |
| Odor | Acrid |
| Refractive Index | 1.383 |
| Quantity | 1 g |
| UN Number | UN3265 |
| Solubility Information | Partly miscible in water. |
| Formula Weight | 92.07 |
| Percent Purity | 96% |
| Show More |
Safety and Handling
| Hazard Category | H302+H312+H332-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H302-H312-H332 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S. |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.