Learn More
2-Fluorotoluene, 99+%
Description
It is employed in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is employed in organic synthesis.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly sealed and store in cool, dry condition in well ventilated place.
Specifications
Specifications
| Melting Point | -87°C |
| Density | 1.002 |
| Boiling Point | 113°C to 114°C |
| Flash Point | 12°C (53°F) |
| Odor | Aromatic |
| Refractive Index | 1.473 |
| Quantity | 2500 g |
| UN Number | UN2388 |
| Beilstein | 1853362 |
| Merck Index | 14,4180 |
| Show More |
Safety and Handling
| Hazard Category | H225-H315-H319-H332-H335 |
| Hazard Statement | GHS H Statement H225-H335 Highly flammable liquid and vapour. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROTOLUENES |
| EINECSNumber | 202-428-5 |
| RTECSNumber | XT2579000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.