Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Formylbenzeneboronic acid, 97%

Click to view available options
Quantity:
1g
5g
40138-16-7
C7H7BO3
149.94
MFCD00151822
DGUWACLYDSWXRZ-UHFFFAOYSA-N
2-formylphenyl boronic acid,2-formylbenzeneboronic acid,benzaldehyde-2-boronic acid,o-formylphenylboronic acid,2-boronobenzaldehyde,2-dihydroxyboryl benzaldehyde,boronic acid, 2-formylphenyl,2-benzaldehyde boronic acid,2-formylphenyl-boronic acid
292189
(2-formylphenyl)boronic acid
B(C1=CC=CC=C1C=O)(O)O
40138-16-7
C7H7BO3
149.94
MFCD00151822
DGUWACLYDSWXRZ-UHFFFAOYSA-N
2-formylphenyl boronic acid,2-formylbenzeneboronic acid,benzaldehyde-2-boronic acid,o-formylphenylboronic acid,2-boronobenzaldehyde,2-dihydroxyboryl benzaldehyde,boronic acid, 2-formylphenyl,2-benzaldehyde boronic acid,2-formylphenyl-boronic acid
292189
(2-formylphenyl)boronic acid
B(C1=CC=CC=C1C=O)(O)O
Applications
2-Formylbenzeneboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Air sensitive. Incompatible with oxidizing agents.
Melting Point ~115°C to 120°C
Quantity 1g
Beilstein 3030776
Formula Weight 149.94
Percent Purity 97%
Chemical Name or Material 2-Formylbenzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning