Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Furoic acid, 98%

Catalog No. AC119825000
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle

CAS: 88-14-2 | C5H4O3 | 112.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-14-2
MDL Number MFCD00003238
InChI Key SMNDYUVBFMFKNZ-UHFFFAOYSA-N
Synonym 2-furoic acid, 2-furancarboxylic acid, pyromucic acid, 2-carboxyfuran, furoic acid, furancarboxylic acid, alpha-furoic acid, alpha-furancarboxylic acid, 2-furanoic acid, kyselina 2-furoova
PubChem CID 6919
ChEBI CHEBI:30845
IUPAC Name furan-2-carboxylic acid
SMILES C1=COC(=C1)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 129.0°C to 133.0°C
Color Beige to White
Boiling Point 230.0°C to 232.0°C
Flash Point 137°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Molecular Formula C5H4O3
Quantity 500 g
Beilstein 18, 272
Merck Index 15, 4336
Solubility Information Solubility in water: 36g/L (20°C). Other solubilities: soluble in alcohol and ether,insoluble in paraffin hydrocarbons
Molecular Weight (g/mol) 112.08
Formula Weight 112.08
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Furoic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
EINECSNumber 201-803-0
RTECSNumber LV1763000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.