Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Furylacetone, 99%
Click to view available options
Quantity:
5 g
25 g
Description
Suitable for fragrance applications.2-Furylacetone is used as the reactant in the synthesis and transformations of furan derivatives
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Suitable for fragrance applications.2-Furylacetone is used as the reactant in the synthesis and transformations of furan derivatives
Solubility
Soluble in alcohol, triacetin.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Suitable for fragrance applications.2-Furylacetone is used as the reactant in the synthesis and transformations of furan derivatives
Solubility
Soluble in alcohol, triacetin.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Boiling Point | 179°C to 180°C |
| Flash Point | 65°C (149°F) |
| Odor | Stench |
| Quantity | 25 g |
| Solubility Information | Soluble in alcohol,triacetin. |
| Formula Weight | 124.14 |
| Percent Purity | 99% |
| Chemical Name or Material | 2-Furylacetone |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 230-234-0 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction