Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Heptyn-1-ol, 97%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
1002-36-4
C7H12O
112.17
MFCD00039541
OGDYROFIIXDTQK-UHFFFAOYSA-N
2-heptyn-1-ol,2-heptyne-1-ol,2-heptynol,pubchem13065,acmc-2097no,hoch2c=c ch2 3ch3
535111
hept-2-yn-1-ol
CCCCC#CCO
1002-36-4
C7H12O
112.17
MFCD00039541
OGDYROFIIXDTQK-UHFFFAOYSA-N
2-heptyn-1-ol,2-heptyne-1-ol,2-heptynol,pubchem13065,acmc-2097no,hoch2c=c ch2 3ch3
535111
hept-2-yn-1-ol
CCCCC#CCO
Boiling Point 89°C to 90°C (16.0mmHg)
Density 0.8800g/mL
Flash Point 78°C
Specific Gravity 0.88
Color Colorless to Yellow
Refractive Index 1.4530 to 1.4550 (20°C, 589nm)
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula CH3(CH2)3C∽CH2OH
Formula Weight 112.17
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Heptyn-1-ol
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning