Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hexadecenoic Acid 98.0+%, TCI America™
SDP

Catalog No. H04281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04281G 1 g
H04285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04281G Supplier TCI America Supplier No. H04281G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 629-56-1
Molecular Formula C16H30O2
Molecular Weight (g/mol) 254.414
MDL Number MFCD00045907
InChI Key ZVRMGCSSSYZGSM-CCEZHUSRSA-N
Synonym Gaidic Acid
PubChem CID 5282743
ChEBI CHEBI:37252
IUPAC Name (E)-hexadec-2-enoic acid
SMILES CCCCCCCCCCCCCC=CC(=O)O

Specifications

Melting Point 49°C
Color White-Yellow
Quantity 1 g
Formula Weight 254.41
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hexadecenoic Acid
EINECSNumber (2)-0609
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.