Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-1-naphthoic Acid 98.0+%, TCI America™
SDP

Catalog No. H0280250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0280250G 250 g
H028025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0280250G Supplier TCI America Supplier No. H0280250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2283-08-1
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.18
MDL Number MFCD00004009
InChI Key UPHOPMSGKZNELG-UHFFFAOYSA-N
Synonym 2-hydroxy-1-naphthoic acid, 1-naphthalenecarboxylic acid, 2-hydroxy, hydroxynaphthoic acid, 1-naphthoic acid, 2-hydroxy, unii-e3h4qz94uy, e3h4qz94uy, 1-naphthalenecarboxylic acid, hydroxy, 2-hydroxynaphthalenecarboxylic acid, 2-hydroxy-1-naphthoicacid, 2-hydroxynaphtoic acid
PubChem CID 16790
IUPAC Name 2-hydroxynaphthalene-1-carboxylic acid
SMILES C1=CC=C2C(=C1)C=CC(=C2C(=O)O)O

Specifications

Melting Point 167°C
Quantity 250 g
Formula Weight 188.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-1-naphthoic Acid
EINECSNumber (4)-0398
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.