Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-2-methylbutyric Acid, 95+%, Spectrum™ Chemical
SDP

Catalog No. 18608221
Click to view available options
Quantity:
5 g
25 g
100 g
25 kg
Packaging:
Poly Bottle

Chemical Identifiers

CAS 3739-30-8
Molecular Weight (g/mol) 118.13 g/mol

Specifications

CAS Max % 1
Color White
Boiling Point 237.6°C
Melting Range 72°C to 75°C
Packaging Poly Bottle
Quantity 100 g
Percent Purity 95%
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material 2-Hydroxy-2-methylbutyric Acid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.