Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Hydroxy-3',4'-dimethoxyacetophenone 98.0+%, TCI America™
SDP

Catalog No. H120525G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 5396-18-9
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.20
MDL Number MFCD00016684
InChI Key BCEPNLMYVYJIHU-UHFFFAOYSA-N
PubChem CID 219639
IUPAC Name 1-(2-hydroxy-3,4-dimethoxyphenyl)ethan-1-one
SMILES COC1=CC=C(C(C)=O)C(O)=C1OC

Specifications

Melting Point 77°C
Quantity 25 g
Formula Weight 196.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Hydroxy-3′,4′-dimethoxyacetophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.