Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone 98.0+%, TCI America™
SDP

Catalog No. H136125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H136125G 25 g
H13615G 5 g
H1361100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H136125G Supplier TCI America Supplier No. H136125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 106797-53-9
Molecular Formula C12H16O4
Molecular Weight (g/mol) 224.26
MDL Number MFCD00085267
InChI Key GJKGAPPUXSSCFI-UHFFFAOYSA-N
Synonym 2-Hydroxy-1-[4-(2-hydroxyethoxy)phenyl]-2-methyl-1-propanone
PubChem CID 86266
IUPAC Name 2-hydroxy-1-[4-(2-hydroxyethoxy)phenyl]-2-methylpropan-1-one
SMILES CC(C)(O)C(=O)C1=CC=C(OCCO)C=C1

Specifications

Melting Point 91°C
Color White-Yellow
Quantity 25 g
Formula Weight 224.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-4′-(2-hydroxyethoxy)-2-methylpropiophenone
EINECSNumber (3)-4272
RTECSNumber UC2976000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.