Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-4-methoxybenzaldehyde, 99%

Catalog No. p-145326
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

Catalog No. AC171080050

Supplier: Thermo Scientific Chemicals 171080050

Only null left
Add to Cart

CAS: 673-22-3 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 673-22-3
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00003327
InChI Key WZUODJNEIXSNEU-UHFFFAOYSA-N
Synonym 4-methoxysalicylaldehyde, benzaldehyde, 2-hydroxy-4-methoxy, 4-methoxysalicyaldehyde, 2-hydroxy-4-methoxy-benzaldehyde, o-hydroxy-p-methoxybenzaldehyde, p-anisaldehyde, 2-hydroxy, salicylaldehyde, 4-methoxy, 2-hydroxy-p-anisaldehyde, 2-formyl-5-methoxyphenol, unii-2n395p88lw
PubChem CID 69600
IUPAC Name 2-hydroxy-4-methoxybenzaldehyde
SMILES COC1=CC=C(C=O)C(O)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 40.0°C to 43.0°C
Color Beige to Cream-White or Brown
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HOC6H3(OCH3)CHO
Quantity 5 g
Beilstein 08, 242
Formula Weight 152.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-4-methoxybenzaldehyde, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-604-0
RTECSNumber BZ2810000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.