Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Hydroxy-5'-methylacetophenone 98.0+%, TCI America™
SDP

Catalog No. H079025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H079025G 25 g
H07905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H079025G Supplier TCI America Supplier No. H079025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1450-72-2
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00002380
InChI Key YNPDFBFVMJNGKZ-UHFFFAOYSA-N
Synonym 2'-hydroxy-5'-methylacetophenone, 1-2-hydroxy-5-methylphenyl ethanone, 2-hydroxy-5-methylacetophenone, o-acetyl-p-cresol, ethanone, 1-2-hydroxy-5-methylphenyl, 2-acetyl-4-methylphenol, 1-2-hydroxy-5-methylphenyl ethan-1-one, 1-hydroxy-2-acetyl-4-methylbenzene, 1-2-hydroxy-5-methyl-phenyl-ethanone, unii-11661u1zen
PubChem CID 15068
IUPAC Name 1-(2-hydroxy-5-methylphenyl)ethanone
SMILES CC1=CC(=C(C=C1)O)C(=O)C

Specifications

Melting Point 47°C
Color Yellow
Boiling Point 114°C
Quantity 25 g
Formula Weight 150.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Hydroxy-5′-methylacetophenone
EINECSNumber (3)-2937
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.