Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-5-methylbenzaldehyde, 99%

Catalog No. p-3593253
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 613-84-3 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-84-3
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00274297
InChI Key ILEIUTCVWLYZOM-UHFFFAOYSA-N
Synonym 5-methylsalicylaldehyde, benzaldehyde, 2-hydroxy-5-methyl, 2,5-cresotaldehyde, 5-methylsalicylic aldehyde, 2-hydroxy-5-methyl-benzaldehyde, 5methylsalicylaldehyde, pubchem15680, 5-methyl salicylaldehyde, 4-hydroxy-m-tolualdehyde, acmc-1bh2k
PubChem CID 69184
IUPAC Name 2-hydroxy-5-methylbenzaldehyde
SMILES CC1=CC=C(O)C(C=O)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 54°C to 57°C
Color Yellow
Boiling Point 217°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H3(OH)CHO
Quantity 1 g
Formula Weight 136.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-5-methylbenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
EINECSNumber 210-357-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.