Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Hydroxy-6'-methoxyacetophenone, 97%

Catalog No. AAA1853906
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1853906 5 g
AAA1853903 1 g
AAA1853914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1853906 Supplier Thermo Scientific Chemicals Supplier No. A1853906
Add to Cart

CAS: 703-23-1 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 703-23-1
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00008732
InChI Key UENLHUMCIOWYQN-UHFFFAOYSA-N
PubChem CID 69709
IUPAC Name 1-(2-hydroxy-6-methoxyphenyl)ethanone
SMILES COC1=CC=CC(O)=C1C(C)=O

Specifications

Melting Point 56°C to 59°C
Boiling Point 141°C (16 mmHg)
Quantity 5 g
Beilstein 1869049
Synonym 1-2-hydroxy-6-methoxyphenyl ethanone, 2'-hydroxy-6'-methoxyacetophenone, 2-hydroxy-6-methoxyacetophenone, 1-2-hydroxy-6-methoxyphenyl ethan-1-one, ethanone, 1-2-hydroxy-6-methoxyphenyl, acetophenone, 2'-hydroxy-6'-methoxy, unii-m5vqg8968n, ethanone,1-2-hydroxy-6-methoxyphenyl, 1-2-hydroxy-6-methoxy-phenyl-ethanone, 1-acetyl-2-hydroxy-6-methoxybenzene
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material 2'-Hydroxy-6'-methoxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-872-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.