Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-6-methylnicotinic acid, 98+%

Catalog No. p-7081741
Click to view available options
Quantity:
5 g
25 g

CAS: 38116-61-9 | C7H7NO3 | 153.137 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38116-61-9
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00006272
InChI Key XRIHTJYXIHOBDQ-UHFFFAOYSA-N
Synonym 2-hydroxy-6-methylnicotinic acid, 2-hydroxy-6-methylpyridine-3-carboxylic acid, 6-methyl-2-oxo-1,2-dihydropyridine-3-carboxylic acid, chembl50725, 3-pyridinecarboxylic acid, 1,2-dihydro-6-methyl-2-oxo, 2-hydroxy-6-methyl nicotinic acid, 2-hydroxy-6-methyl-nicotinic acid, 6-methyl-2-oxo-1,2-dihydro-pyridine-3-carboxylic acid, 6-methyl-2-oxohydropyridine-3-carboxylic acid, 2-hydroxy-6-methylnicotinicacid
PubChem CID 94960
IUPAC Name 6-methyl-2-oxo-1H-pyridine-3-carboxylic acid
SMILES CC1=CC=C(C(=O)N1)C(=O)O

Specifications

Melting Point 233°C to 238°C
Quantity 5 g
Beilstein 129164
Formula Weight 153.14
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxy-6-methylnicotinic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 253-784-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.