Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-9-fluorenone 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
6949-73-1
C13H8O2
196.205
MFCD00001155
GXUBPHMYNSICJC-UHFFFAOYSA-N
2-hydroxy-9-fluorenone,2-hydroxy-9h-fluoren-9-one,2-hydroxy-fluoren-9-one,acmc-209o8z,9h-fluoren-9-one,2-hydroxy,2-hydroxy-9h-fluoren-9-one #,9h-fluoren-9-one, 2-hydroxy
81386
2-hydroxyfluoren-9-one
C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)O
6949-73-1
C13H8O2
196.205
MFCD00001155
GXUBPHMYNSICJC-UHFFFAOYSA-N
2-hydroxy-9-fluorenone,2-hydroxy-9h-fluoren-9-one,2-hydroxy-fluoren-9-one,acmc-209o8z,9h-fluoren-9-one,2-hydroxy,2-hydroxy-9h-fluoren-9-one #,9h-fluoren-9-one, 2-hydroxy
81386
2-hydroxyfluoren-9-one
C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)O
Packaging Glass bottle
Infrared Spectrum Authentic
Color Brown-Red to Pink-Red
Quantity 5g
Melting Point 202°C to 207°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 08, 188
Formula Weight 196.2
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Hydroxy-9-fluorenone, 97%
Show More Show Less

EINECSNumber : 230-119-5

TSCA : TSCA