Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxy-N-methylbenzylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. M10051G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10051G 1 g
M10055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10051G Supplier TCI America Supplier No. M10051G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60399-02-2
Molecular Formula C8H12ClNO
Molecular Weight (g/mol) 173.64
MDL Number MFCD00142934
InChI Key UDXXNWDSBJINMV-UHFFFAOYSA-N
Synonym alpha-Methylamino-o-cresol Hydrochloride, 2-(Methylaminomethyl)phenol Hydrochloride, N-Methyl-2-hydroxybenzylamine Hydrochloride
PubChem CID 44630388
IUPAC Name 2-(methylaminomethyl)phenol;hydrochloride
SMILES CNCC1=CC=CC=C1O.Cl

Specifications

Melting Point 148°C
Quantity 1 g
Formula Weight 173.64
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-N-methylbenzylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.