Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyacetophenone, 97+%

Catalog No. AA4201306
Click to view available options
Quantity:
5 g

CAS: 582-24-1 | C8H8O2 | 136.15 g/mol

2-Hydroxyacetophenone can be used to produce 2-hydroxy-1-phenyl-ethanone-(1)-oxime by heating. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 582-24-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00041829
InChI Key ZWVHTXAYIKBMEE-UHFFFAOYSA-N
Synonym 2-hydroxyacetophenone, benzoylcarbinol, glycolophenone, phenacyl alcohol, alpha-hydroxyacetophenone, 2-hydroxy-1-phenylethan-1-one, ethanone, 2-hydroxy-1-phenyl, methanol, benzoyl, omega-hydroxyacetophenone, acetophenone, 2-hydroxy
PubChem CID 68490
ChEBI CHEBI:28341
IUPAC Name 2-hydroxy-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)CO

Specifications

Melting Point 86°C to 88°C
Quantity 5 g
Sensitivity Air and moisture sensitive
Solubility Information Soluble in water,ether,ethanol,chloroform.
Formula Weight 136.15
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material 2-Hydroxyacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-480-8
TSCA Yes
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.