Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxybenzimidazole, 98%

Catalog No. AAA1832418
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1832418 50 g
AAA1832409 10 g
AAA1832422 100 g
AAA1832430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1832418 Supplier Thermo Scientific Chemicals Supplier No. A1832418
Add to Cart

CAS: 615-16-7 | C7H6N2O | 134.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-16-7
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.14
MDL Number MFCD00127894
InChI Key SILNNFMWIMZVEQ-UHFFFAOYSA-N
Synonym 2-hydroxybenzimidazole, 2-benzimidazolol, 2-benzimidazolinone, o-phenyleneurea, 2-benzimidazolone, 2-oxobenzimidazole, 2 3h-benzimidazolone, 1,3-dihydro-2h-benzimidazol-2-one, 2h-benzimidazol-2-one, 1,3-dihydro, benzamidazole-2 3h-one
PubChem CID 11985
SMILES O=C1NC2=CC=CC=C2N1

Specifications

Melting Point 314°C to 316°C
Quantity 50 g
Beilstein 119866
IUPAC Name 2,3-dihydro-1H-1,3-benzodiazol-2-one
Formula Weight 134.14
Percent Purity 98%
Chemical Name or Material 2-Hydroxybenzimidazole
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 210-412-4
RTECSNumber DE1625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.