Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicone oil, high temperature

Catalog No. AC174665000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle

CAS: 63148-58-3

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63148-58-3
Synonym Poly(methylphenylsiloxane)

Specifications

Density 1.0500g/mL
Flash Point 316°C
Packaging Glass Bottle
Refractive Index 1.4940 to 1.5010
Quantity 500 g
Specific Gravity 1.05
Solubility Information Solubility in water: insoluble in water.
Viscosity 125 mm2/s (25°C)
Chemical Name or Material Silicone oil

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.