Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxycyclohexanone, dimer, 90+%

Catalog No. p-3598167
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 30282-14-5 | C12H20O4 | 228.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30282-14-5
Molecular Formula C12H20O4
Molecular Weight (g/mol) 228.29
MDL Number MFCD00151157
InChI Key ODZTXUXIYGJLMC-UHFFFAOYSA-N
Synonym 2-hydroxycyclohexanone, adipoin, 2-hydroxy-1-cyclohexanone, cyclohexanone, 2-hydroxy, 2-hydroxy-cyclohexanone, cyclohexanone, 2-hydroxy-, 2r, cyclohexanone, 2-hydroxy-, dimer, cyclohexanone, dimer, zlchem 1128, acmc-20n5uh
PubChem CID 10785
ChEBI CHEBI:17878
IUPAC Name 2-hydroxycyclohexan-1-one
SMILES C1CCC(=O)C(C1)O

Specifications

CAS Min % 90.0
CAS Max % 100.0
Melting Point 100°C to 107°C
Color White
Boiling Point 83°C to 86°C (13.0 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 228.29
Percent Purity 90+%
Physical Form Powder
Chemical Name or Material 2-Hydroxycyclohexanone, dimer
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable solid.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.