Learn More
2-Hydroxyethyl acrylate, 97%, stab. with 200-300ppm 4-methoxyphenol
Description
As a reactive monomer2-Hydroxyethyl acrylate is used as a crosslinking agent for resins, plastics and rubber modifiers. Further, it is used in the synthesis of amphilic block copolymers by nitroxide mediated living radical polymerization. In addition to this, it is used to prepare tuned poly(hydroxyethyl acrylate) by atom transfer radical polymerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
As a reactive monomer2-Hydroxyethyl acrylate is used as a crosslinking agent for resins, plastics and rubber modifiers. Further, it is used in the synthesis of amphilic block copolymers by nitroxide mediated living radical polymerization. In addition to this, it is used to prepare tuned poly(hydroxyethyl acrylate) by atom transfer radical polymerization.
Solubility
Miscible with water.
Notes
Light sensitive. Incompatible with acids, bases, peroxides, metals, copper and oxidizing agents.

Specifications
Specifications
Melting Point | <-60°C |
Density | 1.106 |
Boiling Point | 192°C |
Flash Point | 101°C (214°F) |
Odor | Pungent |
Refractive Index | 1.45 |
Linear Formula | CH2=CHCO2CH2CH2OH |
Quantity | 1000 g |
UN Number | UN2927 |
Sensitivity | Light sensitive |
Show More |
Safety and Handling
Hazard Category | H302-H310-H314-H317 |
Hazard Statement | GHS H Statement H311-H314-H317 Toxic in contact with skin. Causes severe skin burns and eye damage. May cause an allergic skin reaction. |
Precautionary Statement | P260-P262-P264b-P270-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P362-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S. |
EINECSNumber | 212-454-9 |
RTECSNumber | AT1750000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.