Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Hydroxyethyl acrylate, 97%, stab. with 200-300ppm 4-methoxyphenol

Click to view available options
Quantity:
250g
1000g
818-61-1
C5H8O3
116.116
MFCD00002865
OMIGHNLMNHATMP-UHFFFAOYSA-N
2-hydroxyethyl acrylate,hydroxyethyl acrylate,2-propenoic acid, 2-hydroxyethyl ester,ethylene glycol monoacrylate,bisomer 2hea,acrylic acid 2-hydroxyethyl ester,2-acryloyloxy ethanol,ethylene glycol, acrylate,acrylic acid, 2-hydroxyethyl ester,2-hydroxyethylacrylate
13165
2-hydroxyethyl prop-2-enoate
C=CC(=O)OCCO
818-61-1
C5H8O3
116.116
MFCD00002865
OMIGHNLMNHATMP-UHFFFAOYSA-N
2-hydroxyethyl acrylate,hydroxyethyl acrylate,2-propenoic acid, 2-hydroxyethyl ester,ethylene glycol monoacrylate,bisomer 2hea,acrylic acid 2-hydroxyethyl ester,2-acryloyloxy ethanol,ethylene glycol, acrylate,acrylic acid, 2-hydroxyethyl ester,2-hydroxyethylacrylate
13165
2-hydroxyethyl prop-2-enoate
C=CC(=O)OCCO
Applications
As a reactive monomer2-Hydroxyethyl acrylate is used as a crosslinking agent for resins, plastics and rubber modifiers. Further, it is used in the synthesis of amphilic block copolymers by nitroxide mediated living radical polymerization. In addition to this, it is used to prepare tuned poly(hydroxyethyl acrylate) by atom transfer radical polymerization.

Solubility
Miscible with water.

Notes
Light sensitive. Incompatible with acids, bases, peroxides, metals, copper and oxidizing agents.
Odor Pungent
Melting Point <-60°C
Quantity 250g
Boiling Point 192°C
Flash Point 101°C (214°F)
Linear Formula CH2=CHCO2CH2CH2OH
UN Number UN2927
Formula Weight 116.12
Percent Purity 97%
Chemical Name or Material 2-Hydroxyethyl acrylate, stabilized with 200 to 300ppm 4-methoxyphenol

GHS H Statement
H311-H314-H317
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.

RTECSNumber : AT1750000

TSCA : TSCA