Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Hydroxyethyl Disulfide 95%, Thermo Scientific™

Catalog No. AC176241000
Click to view available options
Quantity:
25 g
100 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1892-29-1
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00002906
InChI Key KYNFOMQIXZUKRK-UHFFFAOYSA-N
Synonym 2-hydroxyethyl disulfide, 2,2'-dithiodiethanol, bis 2-hydroxyethyl disulfide, ethanol, 2,2'-dithiobis, hydroxyethyl disulfide, diethanol disulfide, 2,2'-dithiobisethanol, 2-hydroxyethanedisulfide, usaf th-9, 2,2'-disulfanediyldiethanol
PubChem CID 15906
ChEBI CHEBI:43136
IUPAC Name 2-[(2-hydroxyethyl)disulfanyl]ethan-1-ol
SMILES OCCSSCCO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Density 1.26
Infrared Spectrum Authentic
Refractive Index 1.5650 to 1.5710 (20°C, 589nm)
Linear Formula (HOCH2CH2)2S2
Quantity 100 g
Beilstein 01, 471
Formula Weight 154.25
Percent Purity ≥94%
Physical Form Viscous Liquid After Melting
Chemical Name or Material 2-Hydroxyethyl disulfide, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 217-576-6
RTECSNumber KK7525000
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.